missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic Acid (Certified ACS), Fisher BioReagents

Catalog Number p-27342
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
139-13-9
C6H9NO6
191.14
MFCD00004287
MGFYIUFZLHCRTH-UHFFFAOYSA-N
nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
8758
CHEBI:44557
2-[bis(carboxymethyl)amino]acetic acid
OC(=O)CN(CC(O)=O)CC(O)=O
This item is not returnable. View return policy
139-13-9
C6H9NO6
191.14
MFCD00004287
MGFYIUFZLHCRTH-UHFFFAOYSA-N
nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
8758
CHEBI:44557
2-[bis(carboxymethyl)amino]acetic acid
OC(=O)CN(CC(O)=O)CC(O)=O
Nitrilotriacetic Acid is a chelating and sequestering agent. A builder in synthetic detergents.
TRUSTED_SUSTAINABILITY
Melting Point 245°C
Color White
pH 2.3
Assay Percent Range ≥98.0 %
Packaging Poly Bottle
Linear Formula N(CH2COOH)3
Quantity 100 g
Merck Index 15, 6665
Formula Weight 191.14
Percent Purity ≥98.0%
Grade Certified ACS
Physical Form Powder or Solid
Chemical Name or Material Nitrilotriacetic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitrilotriacetic acid
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'