missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-172536
14.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy
139-13-9
C6H9NO6
191.14
MFCD00004287
MGFYIUFZLHCRTH-UHFFFAOYSA-N
nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
8758
CHEBI:44557
2-[bis(carboxymethyl)amino]acetic acid
OC(=O)CN(CC(O)=O)CC(O)=O
This item is not returnable. View return policy
139-13-9
C6H9NO6
191.14
MFCD00004287
MGFYIUFZLHCRTH-UHFFFAOYSA-N
nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
8758
CHEBI:44557
2-[bis(carboxymethyl)amino]acetic acid
OC(=O)CN(CC(O)=O)CC(O)=O

Nitrilotriacetic acid, CAS # 139-13-9, is a tricarboxylic acid compound used for chemical chelation and elution in protein studies and the purification of certain earth compounds.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Nitrilotriacetic acid (2,2',2-nitrilotriacetic acid) is a tricarboxylic acid used for chemical purification


Applications

  • A known mutagen, suitable for cell culture experiments of DNA damage
  • Nitrilotriacetic acid can be used for protein elution using the His-tag method and chemical chelation
  • Nitrilotriacetic acid is capable of chelating Ca2+, Co2+, Cu2+, and Fe3+
  • Intended for sensitive applications such as HPLC
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 245.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula N(CH2CO2H)3
Quantity 5 g
Beilstein 04,369
Merck Index 15,6665
Solubility Information Solubility in water: 1.28g/L (22.5°C). Other solubilities: soluble in most common organic solvents
Formula Weight 191.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Nitrilotriacetic acid, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitrilotriacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'