Learn More
Kimberly-Clark™ KLEENGUARD™ G20 Nitrile Gloves
Kimberly-Clark™ KLEENGUARD™ G20 Nitrile Gloves are an ambidextrous glove that provides protection against chemical splash in a wide range of applications.
Brand: Kimberly-Clark™ 90092
Code : NEW
Additional Details : Weight : 11.07000kg
Description
- PPE Category 3 protection
- For increased protection against chemical splashes of several chemicals.
- Provides exceptional touch sensitivity and textured fingertips to improve grip and the handling of small objects
- AQL 0.65 for pinholes guarantees quality of production process, waste reduction and better protection
- Independently accredited
- Suitable for food handling
- Beaded cuff for added strength and ease of donning
- Latex Free
- Powder free minimises srying effects of pwder and skin allergies
- Ambidextrous
- Portable dispenser box for mobility and easy access
Specifications
Green | |
Nitrile | |
Chemical | |
EN374-3: 2003, EN374-2:2003, EN388:2003 |
Long, beaded | |
2500/Pk. | |
Medium |