Learn More
Meditrade™ NITRIL™ BestGen™ Disposable Nitrile Examination Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Description
- Medical glove tested according to DIN EN 455
- PPE according to EN 374
- Protects against dangerous chemicals and micro-organisms
- Suitable for food use
- Ideal for users with sensitive skin with diagnosed latex allergy (Type I)
- Latex free
- Disposable
- Touch feeling formula offers excellent touch and gives it an almost latex-like high elasticity
- Tear resistant
- Polymer inner coating makes them easy to put on
- Ambidextrous with roll edge
- Blue
- Powder-free
- Micro-rough
- Non-sterile
- Single-use
Specifications
Certifications/Compliance (Europe) | EN ISO 13485:2016; 93/42/EWG/EEC Class I Medical Product; (EU) 2016/425 PPE Category III; DIN EN 455-1/ -2/ -3/ -4; EN 374-1/ -2/ -4/ -5; EN 16523-1; EN 420 |
Color | Blue |
Disposable | Single-use |
Latex Free | Yes |
Material | Nitrile |
Size | X-Small |