Learn More
Nitric Acid 67-69%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
HNO3, CAS Number-7697-37-2, acidum nitricum, acide nitrique, aqua fortis, azotic acid, salpetersaeure, hydrogen nitrate, nitryl hydroxide, rfna, nital, nitrous fumes, 1L, CHEBI:48107, Colorless Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, Liquid, 944
Brand: Fisher Chemical A509-P1
205.86 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 25
Additional Details : CAS Number : 7697-37-2 Weight : 1.40000kg Transport : UN number : 2031 Chem class : 8 Pack group : II
Description
HNO3, CAS Number-7697-37-2, acidum nitricum, acide nitrique, aqua fortis, azotic acid, salpetersaeure, hydrogen nitrate, nitryl hydroxide, rfna, nital, nitrous fumes, 1L, CHEBI:48107, Colorless Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, Liquid, 944
PolybottleSpecifications
1 | |
-41°C | |
0.94kPa (20°C) | |
HNO3 | |
MFCD00011349 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.02g/mol |
Colorless Yellow | |
7697-37-2 | |
1 L | |
Liquid | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 |
Safety and Handling
- Nitric acid 67-69%
Signal Word
- Danger
Hazard Category
- Oxidising liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Nitric acid