missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™

Catalog Number 11974091
468.13 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
FEP Bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O

Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL

  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially manufactured and cleaned FEP fluoropolymer bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.
TRUSTED_SUSTAINABILITY
pH 1
Color Colorless Yellow
Melting Point -41°C
Boiling Point 120.5°C
Density 1.40 g/cm3
Packaging FEP Bottle
Vapor Pressure 0.94kPa (20°C)
Certificate of Analysis Certificate of Lot Analysis is included with each bottle.
Quantity 250 mL
Physical Form Liquid
UN Number 2031
Merck Index 15, 6663
Formula Weight 63.01
Grade OPTIMA™
Chemical Name or Material Nitric Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitric acid 67-69%, Optima for Ultra traces analysis
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'