missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric acid, 70%, in water, Thermo Scientific™
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Appearance | Clear liquid (may darken on storage) |
Linear Formula | HNO3 |
UN Number | 2031 |
Fieser | 05-474 |
Merck Index | 6608 |
Composition | 70% Nitric acid, 30% water |
Concentration | 67% to 70% |
Density | 1.4 |
Formula Weight | 63.01 |
Physical Form | Liquid |
Show More |