missing translation for 'onlineSavingsMsg'
Learn More

Nitinol wire, 1mm, Alfa Aesar™

Form: approx. 49.71g/10m; Af approx. 10°C. Solid in various forms

Brand:  Alfa Aesar 045087.H5

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150m

 View more versions of this product

Product Code. 15443945

  • £1098.00 / 150m


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Nitinol
Wire, 1.0mm (0.039 in.) dia.
MFCD02091734

Specifications

Nitinol
150m
Odorless
MFCD02091734
Silver-gray
Silver gray wire
Wire, 1.0mm (0.039 in.) dia.
Videos
SDS