missing translation for 'onlineSavingsMsg'
Learn More

Nitinol wire, , 0.58mmAlfa Aesar™

Form: approx. 17.29g/10m; Af approx. 10°C; Superelastic; Oxide surface

Brand:  Alfa Aesar 044951.H2

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 50m

 View more versions of this product

Product Code. 11420940

  • £308.10 / 50m


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Chemical Identifiers

Nitinol
Alloy N, straight annealed
Wire, 0.58mm (0.023 in.) dia.
MFCD02091734

Specifications

Nitinol
50m
Silver-gray wire
Wire, 0.58mm (0.023 in.) dia.
Silver-gray
Alloy N, straight annealed
Odorless
MFCD02091734
Videos
SDS