missing translation for 'onlineSavingsMsg'
Learn More

Nitinol wire, , 0.3mmAlfa Aesar™

Brand:  Alfa Aesar 044948.H5

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150m

 View more versions of this product

Product Code. 11440930

  • £694.40 / 150m


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Nitinol wire
∽5.52g/10m; Af approx. 10°C
MFCD02091734

Specifications

Nitinol wire
150m
∽5.52g/10m; Af approx. 10°C
MFCD02091734
Videos
SDS