missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Nitinol tubing, 0.7mm (0.028in) OD, 0.58mm (0.023in) ID, superelastic, Alloy S, straight annealed, oxide surface
Brand: Alfa Aesar™ 045857.BZ
Code : NEW
Additional Details : Weight : 0.01000kg
Quantity | 150cm |
---|
Chemical Identifiers
Nitinol tubing | |
OD: 0.7mm (0.028 in.), ID: 0.58mm (0.023 in.), Superelastic, Alloy S, Straight annealed, Oxide surface | |
MFCD02091734 |
Specifications
Nitinol tubing | |
OD: 0.7mm (0.028 in.), ID: 0.58mm (0.023 in.), Superelastic, Alloy S, Straight annealed, Oxide surface | |
MFCD02091734 |
150cm | |
Odorless |