Learn More
Nitinol foil, 0.127mm (0.005in) thick, shape memory, flat annealed, pickled surface, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045514.GH
609.36 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Nitinol foil is used in cell phone antennas, children's toys, experimental materials, eye glasses components and medical equipment. It is also used in heat engines, resilient glass frames, mechanical watch springs, temperature control system, couplings, golf club insert, biomedical and medical uses, cell-phone technology: retractable antenna, microphone boom. It is highly biocompatible and has properties suitable for use in orthopaedic implants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nitinol foil | |
MFCD02091734 | |
Insoluble in water. | |
≈0.51g/25x25mm |
0.127mm (0.005 in.) thick, shape memory, flat annealed, pickled surface | |
100 x 100 mm | |
Odorless |
RUO – Research Use Only