missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nitinol foil, 0.127mm (0.005 in.) thick, shape memory, flat annealed, pickled surface

Brand:  Alfa Aesar™ 045514.FI

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 50 x 50mm

 View more versions of this product

Product Code. 15433995

  • £267.40 / Each


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

Nitinol foil
0.127mm (0.005 in.) thick, shape memory, flat annealed, pickled surface
MFCD02091734

Specifications

Nitinol foil
50 x 50mm
Odorless
MFCD02091734
≈0.51g/25x25mm
0.127mm (0.005 in.) thick, shape memory, flat annealed, pickled surface
Af approx. 45°C
Videos
SDS