missing translation for 'onlineSavingsMsg'
Learn More

Nitinol compression coil spring, wire diameter 1.0mm (0.039in), mean coil diameter 7.0mm (0.27in), Alloy M, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 045578.KM

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15483995

  • £272.00 / Pack of 25
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Nitinol compression coil spring, Alloy M
MFCD02091734
Odorless
Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.)
25 Pc.
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers