Learn More
Thermo Scientific Alfa Aesar NIST traceable size standard, polystyrene monodisperse, 30µm, 1% wt%, aqueous suspension in dropper-tipped bottles
Brand: Thermo Scientific Alfa Aesar J67759.4Y
NIST traceable size standard, polystyrene monodisperse, 30µm, 1% wt%, aqueous suspension in dropper-tipped bottles
Code : NEW
Additional Details : CAS Number : 9003-53-6 Weight : 0.00015kg