missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ NIST traceable size standard, polystyrene monodisperse, 2μm, 1% wt%, aqueous suspension in dropper-tipped bottles

Brand:  Alfa Aesar™ J67531.4Y

Code : NEW

Additional Details : CAS Number : 9003-53-6 Weight : 0.00015kg

Product Code. 15438759

  • £263.94 / 15mL


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Specifications

NIST traceable size standard, polystyrene monodisperse, 2μm
9003-53-6
Liquid
dropper-tipped bottles
15mL
1% wt%, aqueous suspension
1.050g/mL
Odorless
White
Videos
SDS