Learn More
Nisoxetine hydrochloride, Tocris Bioscience™
Noradrenalin re-uptake inhibitor
Brand:  Tocris (Bio-Techne) 102550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13468323
  • £382.20 / 50mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
A potent and selective inhibitor of noradrenalin uptake with little or no affinity for a range of other neurotransmitter receptors.
Specifications
Chemical Identifiers
57754-86-6
307.818
hydrogen nisoxetine chloride
3-(2-methoxyphenoxy)-N-methyl-3-phenylpropan-1-amine;hydrochloride
C17H22ClNO2
LCEURBZEQJZUPV-UHFFFAOYSA-N
134453
CNCCC(C1=CC=CC=C1)OC2=CC=CC=C2OC.Cl
Specifications
Nisoxetine hydrochloride
57754-86-6
hydrogen nisoxetine chloride
CNCCC(C1=CC=CC=C1)OC2=CC=CC=C2OC.Cl
307.818
307.82
50mg
C17H22ClNO2
LCEURBZEQJZUPV-UHFFFAOYSA-N
3-(2-methoxyphenoxy)-N-methyl-3-phenylpropan-1-amine;hydrochloride
134453
>99%
Videos
Documentos