missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIPSNAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563688
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EYLDAYNSLT EAVLPKLHLD EDYPCSLV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
NIPSNAP1 Polyclonal antibody specifically detects NIPSNAP1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
NIPSNAP1 | |
Polyclonal | |
Unconjugated | |
NIPSNAP1 | |
4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 1; 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1; 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1 (C. elegans); nipsnap homolog 1 (C. elegans); Nipsnap1; Protein NipSnap homolog 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8508 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BPW8 | |
NIPSNAP1 | |
Recombinant protein corresponding to Human NIPSNAP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only