missing translation for 'onlineSavingsMsg'
Learn More

Niobium powder, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number 15465935
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
This item is not returnable. View return policy
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
23936
CHEBI:33344
niobium
[Nb]
This item is not returnable. View return policy
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
23936
CHEBI:33344
niobium
[Nb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3089
Quantity 2.5 g
Percent Purity 99.8%
Mesh Size −325 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Niobium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'