missing translation for 'onlineSavingsMsg'
Learn More

Niobium potassium n-butoxide, 5% w/v in n-butanol, 99% (metals basis), Thermo Scientific™

Catalog Number p-7059221
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
C24H54KNbO6
MFCD02682834
This item is not returnable. View return policy
C24H54KNbO6
MFCD02682834

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Linear Formula KNb(OC4H9)6
Quantity 25 mL
UN Number UN1120
Sensitivity Moisture sensitive
Formula Weight 570.01
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 5% w/v in n-butanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Niobium potassium n-butoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'