missing translation for 'onlineSavingsMsg'
Learn More

Niobium potassium isopropoxide, 2.5% w/v in isopropanol, 99% (metals basis), Thermo Scientific™

Catalog Number p-7050635
Click to view available options
Quantity:
50 mL
200 mL
This item is not returnable. View return policy
21864-23-3
C12H31KNbO4
371.381
MFCD00798516
WLSOPSYOUMIABE-UHFFFAOYSA-N
niobium potassium isopropoxide w/v in isopropanol
121233692
potassium;niobium;propan-2-ol;propan-2-olate
CC(C)O.CC(C)O.CC(C)O.CC(C)[O-].[K+].[Nb]
This item is not returnable. View return policy
21864-23-3
C12H31KNbO4
371.381
MFCD00798516
WLSOPSYOUMIABE-UHFFFAOYSA-N
niobium potassium isopropoxide w/v in isopropanol
121233692
potassium;niobium;propan-2-ol;propan-2-olate
CC(C)O.CC(C)O.CC(C)O.CC(C)[O-].[K+].[Nb]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Brown to Yellow
Flash Point 12°C (54°F)
Odor Alcohol-like
Linear Formula KNb[OCH(CH3)2]6
Quantity 50 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 486.53
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 2.5% w/v in isopropanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Niobium potassium isopropoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'