missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Niobium n-propoxide, 99% (metals basis)

Brand:  Alfa Aesar™ 036571.06

Code : NEW

Additional Details : CAS Number : 38874-17-8 Weight : 0.00500kg Transport : UN number : 1993 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11387998

  • £136.80 / 5g


Estimated Shipment date
from Supplier 23-04-2018
Add to basket

Specifications

Chemical Identifiers

Niobium n-propoxide
Nb(OC3H7)5
MFCD00144917
38874-17-8
388.35
UN1993

Specifications

Niobium n-propoxide
166°C (0.05mmHg)
38874-17-8
99%
Nb(OC3H7)5
MFCD00144917
Moisture Sensitive
5g
Liquid
(metals basis)
388.35
UN1993
Videos
SDS