missing translation for 'onlineSavingsMsg'
Learn More

Niobium n-pentyloxide, 99% (metals basis), Thermo Scientific™

Catalog Number p-7050345
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
105091-67-6
MFCD00144918
This item is not returnable. View return policy
105091-67-6
MFCD00144918

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Linear Formula Nb(OC5H11)5
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 528.18
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Niobium n-pentyloxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'