missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niobium isopropoxide, 99% (metals basis), 10% w/v in isopropanol/hexane (50:50), Thermo Scientific Chemicals
82.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 mL
100 mL
Description
Niobium isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Linear Formula | Nb[OCH(CH3)2]5 |
Quantity | 25 mL |
UN Number | UN1993 |
Sensitivity | Moisture sensitive |
Solubility Information | It reacts with water. Soluble in hydrocarbons and warm isopropanol,reacts with other alcohols,ketones and esters. |
Formula Weight | 388.35 |
Percent Purity | 99% |
Concentration or Composition (by Analyte or Components) | 10% w/v in isopropanol/hexane (50:50) |
Physical Form | Liquid |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'