missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nile Blue A

Catalog Number p-7023528
15.68 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3625-57-8
C40H40N6O6S
732.84
MFCD00064529
QIRDPEPUXNCOLD-UHFFFAOYSA-N
19256
[9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]
This item is not returnable. View return policy
3625-57-8
C40H40N6O6S
732.84
MFCD00064529
QIRDPEPUXNCOLD-UHFFFAOYSA-N
19256
[9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]

Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.

Solubility
Soluble in ethanol, and water (50 g/L) at 25°C.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point >300°C (decomposition)
Synonym C.I. 51180, Basic Blue 12
Solubility Information Soluble in ethanol,and water (50g/L) at 25°C.
Formula Weight 732.84
Quantity 5 g
Chemical Name or Material Nile Blue A

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'