missing translation for 'onlineSavingsMsg'
Learn More

Nifedipine, ACROS Organics™

£39.90 - £311.90

Chemical Identifiers

CAS 21829-25-4
Molecular Formula C17H18N2O6
Molecular Weight (g/mol) 346.3
MDL Number MFCD00057326
InChI Key HYIMSNHJOBLJNT-UHFFFAOYSA-N
Synonym nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord
PubChem CID 4485
ChEBI CHEBI:7565
IUPAC Name dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CC1=C(C(C(=C(N1)C)C(=O)OC)C2=CC=CC=C2[N+](=O)[O-])C(=O)OC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10405795 SDS View Documents Acros Organics
329270010
1g Glass bottle £39.90
1g View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
10365991 SDS View Documents Acros Organics
329270050
5g Glass bottle £98.00
5g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
10036213 SDS View Documents Acros Organics
329270250
25g Glass bottle £311.90
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 11-06-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

21829-25-4
346.3
HYIMSNHJOBLJNT-UHFFFAOYSA-N
4485
dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
C17H18N2O6
MFCD00057326
nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord
CHEBI:7565
CC1=C(C(C(=C(N1)C)C(=O)OC)C2=CC=CC=C2[N+](=O)[O-])C(=O)OC

Specifications

Nifedipine
Authentic
Glass bottle
White to Yellow
1g
C17H18N2O6
22, V,4, 268
nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord
HYIMSNHJOBLJNT-UHFFFAOYSA-N
dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
4485
346.3
98.0 to 102.0% (on dried substance)
0.1% max.
0.5% max.
0.1% max
171.0°C to 175.0°C
21829-25-4
MFCD00057326
15, 6613
Solubility in water: insoluble in water. Other solubilities: soluble in acetone,chloroform,benzene,dioxane,,methanol,ethanol
CC1=C(C(C(=C(N1)C)C(=O)OC)C2=CC=CC=C2[N+](=O)[O-])C(=O)OC
346.3
CHEBI:7565
Powder
Safety and Handling

Safety and Handling

Product Identifier
  • Nifedipine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.