missing translation for 'onlineSavingsMsg'
Learn More

NIDA 41020, Tocris Bioscience™

High affinity CB1 antagonist

Brand:  Tocris (Bio-Techne) 392110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13463814

  • £151.41 / 10mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity CB1 receptor antagonist (Ki = 4.1 nM). Exhibits significantly reduced lipophilicity compared to other CB1 antagonists.
Specifications

Chemical Identifiers

502486-89-7
459.371
unii-83e62l67id, 1-2,4-dichlorophenyl-5-4-methoxyphenyl-4-methyl-n-1-piperidinyl-1h-pyrazole-3-carboxamide, n-piperidin-1-yl-5-4-methoxyphenyl-1-2,4-dichlorophenyl-4-methyl-1h-pyrazole-3-carboxamide, nida hplc , solid, 1-2,4-dichloro-phenyl-5-4-methoxy-phenyl-4-methyl-1h-pyrazole-3-carboxylic acid piperidin-1-ylamide, 1-2,4-dichlorophenyl-5-4-methoxyphenyl-4-methyl-n-piperidin-1-yl pyrazole-3-carboxamide, 1h-pyrazole-3-carboxamide, 1-2,4-dichlorophenyl-5-4-methoxyphenyl-4-methyl-n-1-piperidinyl
1-(2,4-dichlorophenyl)-5-(4-methoxyphenyl)-4-methyl-N-piperidin-1-ylpyrazole-3-carboxamide
C23H24Cl2N4O2
KWDBQJRWPWTGPF-UHFFFAOYSA-N
11037861
CC1=C(N(N=C1C(=O)NN2CCCCC2)C3=C(C=C(C=C3)Cl)Cl)C4=CC=C(C=C4)OC

Specifications

NIDA 41020
502486-89-7
unii-83e62l67id, 1-2,4-dichlorophenyl-5-4-methoxyphenyl-4-methyl-n-1-piperidinyl-1h-pyrazole-3-carboxamide, n-piperidin-1-yl-5-4-methoxyphenyl-1-2,4-dichlorophenyl-4-methyl-1h-pyrazole-3-carboxamide, nida hplc , solid, 1-2,4-dichloro-phenyl-5-4-methoxy-phenyl-4-methyl-1h-pyrazole-3-carboxylic acid piperidin-1-ylamide, 1-2,4-dichlorophenyl-5-4-methoxyphenyl-4-methyl-n-piperidin-1-yl pyrazole-3-carboxamide, 1h-pyrazole-3-carboxamide, 1-2,4-dichlorophenyl-5-4-methoxyphenyl-4-methyl-n-1-piperidinyl
CC1=C(N(N=C1C(=O)NN2CCCCC2)C3=C(C=C(C=C3)Cl)Cl)C4=CC=C(C=C4)OC
459.371
459.37
10mg
C23H24Cl2N4O2
KWDBQJRWPWTGPF-UHFFFAOYSA-N
1-(2,4-dichlorophenyl)-5-(4-methoxyphenyl)-4-methyl-N-piperidin-1-ylpyrazole-3-carboxamide
11037861
>99%
Videos
Documents