missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic acid, specified according to the requirements of Ph. Eur, Thermo Scientific Chemicals

Catalog Number p-3361489
40.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
59-67-6
C6H5NO2
123.11
MFCD00006391
PVNIIMVLHYAWGP-UHFFFAOYSA-N
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
938
CHEBI:15940
pyridine-3-carboxylic acid
OC(=O)C1=CC=CN=C1
59-67-6
C6H5NO2
123.11
MFCD00006391
PVNIIMVLHYAWGP-UHFFFAOYSA-N
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
938
CHEBI:15940
pyridine-3-carboxylic acid
OC(=O)C1=CC=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 236.0°C to 239.0°C
Packaging Plastic bottle
Quantity 500 g
Merck Index 15,661
Solubility Information (in water) Sparingly soluble
Formula Weight 123.11
Related Substances Passes test
Grade Specified according to the requirements of Ph. Eur
Chemical Name or Material Nicotinic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'