missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Nicotine Tartrate (White Solid), Fisher BioReagents
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass
Description
(-)-Nicotine Tartrate is used in biochemical research applications.
Specifications
Melting Point | 98.00°C |
Color | White |
Packaging | Amber Glass |
Quantity | 1 g |
Merck Index | 15, 6609 |
Specific Rotation | Report ° (+ or -) |
Formula Weight | 462.4 |
Physical Form | Solid |
Chemical Name or Material | (-)-Nicotine Tartrate |
Safety and Handling
Product Identifier
- (-)-Nicotine Tartrate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- H300-Fatal if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'