missing translation for 'onlineSavingsMsg'
Learn More

Nicotine Bitartrate Dihydrate, MP Biomedicals™

Catalog Number 11492902
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
C18H30N2O14
498.44
MFCD00150899
LDMPZNTVIGIREC-HYRPJEHINA-N
22464
O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1
This item is not returnable. View return policy
C18H30N2O14
498.44
MFCD00150899
LDMPZNTVIGIREC-HYRPJEHINA-N
22464
O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1
CAS 1504589
Melting Point 93°C to 95°C (literature)
Color White
pH 3.18 (10% aq. soln.) (literature)
Quantity 1 g
Synonym 4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d
Solubility Information Soluble in water (50mg/mL - clear to slightly hazy, colorless to faint yellow solution) or alcohol
IUPAC Name pent-4-ynoic acid
Formula Weight 498.44
Physical Form Powder
Chemical Name or Material Nicotine Bitartrate Dihydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nicotine Bitartrate Dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Nicotine BitartrateDihydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'