missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 98%, MP Biomedicals™
Click to view available options
Quantity:
25 mg
50 mg
1 g
Specifications
Color | White |
Quantity | 25 mg |
IUPAC Name | [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |
Formula Weight | 833.4 |
Percent Purity | 0.98 |
Physical Form | Powder |
Chemical Name or Material | β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'