missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel Vanadium, sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

Brand:  Alfa Aesar™ 040881.KS

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15413745

  • £395.38 / Each


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Specifications

Ni:V; 93:7 wt%
1 each
Non-Magnetic
(metals basis)
Silver/gray
Nickel Vanadium, sputtering target
76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
99.95%
MFCD01310520
Videos
SDS