missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel Ultrathin foil, 1.0 micron thick, 99.99+% (metals basis)

Brand:  Alfa Aesar™ 040694.HF

Code : NEW

Additional Details : CAS Number : 7440-02-0 Weight : 0.01000kg

Quantity 10mm

 View more versions of this product

Product Code. 11388628

  • £377.10 / Each


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Nickel Ultrathin foil
7440-02-0
MFCD00011137

Specifications

Nickel Ultrathin foil
7440-02-0
≥99.99%
MFCD00011137
1.0 Micron thick
10mm
Circular Disc on Permanent Mylar Support
(metals basis)
Odorless
Videos
SDS