Learn More
Nickel Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical
Brand: Fisher Chemical J/8055/08
Code : 23
Additional Details : CAS Number : 7440-02-0 Weight : 0.50000kg Transport : UN number : 2031 Chem class : 8 Pack group : II
Quantity | 500mL |
---|
Chemical Identifiers
11137 |
Specifications
11137 | |
HDPE plastic bottle | |
1 |
Liquid | |
Colorless | |
500mL |