missing translation for 'onlineSavingsMsg'
Learn More

Nickel rod, 12mm (0.47in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number 11317177
Click to view available options
Quantity:
10 mm
50 mm
150 mm
This item is not returnable. View return policy
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]
This item is not returnable. View return policy
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 mm
Percent Purity 99.995%
Grade Puratronic™
Odor Odorless
Weight ≈10.1g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Nickel rod, 12mm (0.47 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'