missing translation for 'onlineSavingsMsg'
Learn More

Nickel powder, 99.996% (metals basis), Alfa Aesar™, Puratronic™

C typically less-than0.05%

Brand:  Alfa Aesar 010674.14

Code : NEW

Additional Details : CAS Number : 7440-02-0 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15404745

  • £54.70 / 25g


Estimated Shipment date
from Supplier 22-01-2018
Add to basket

Specifications

Chemical Identifiers

Nickel
Powder
MFCD00011137
7440-02-0
99.996%

Specifications

Nickel
7440-02-0
8.908g/cm3 at 20°C
99.996%
Odorless
25g
Silver gray powder
Powder
MFCD00011137
Silver-gray
Videos
SDS