Learn More
Nickel plating solution, electroless, ammonia free, Thermo Scientific Chemicals
For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar
Brand: Thermo Scientific Alfa Aesar 044070.AK
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Green | |
Electroless, ammonia free | |
UN3082 |
Nickel plating solution | |
MFCD00011137 | |
250 mL |
RUO – Research Use Only