missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel, plasma standard solution, Specpure™, Ni 10μg/mL

Brand:  Alfa Aesar™ 045261.AE

Code : NEW

Additional Details : Weight : 0.00010kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 100mL

Product Code. 15406865

  • £40.40 / 100mL


Estimated Shipment date
from Supplier 22-06-2018
Add to basket

Specifications

Specifications

Nickel, plasma standard solution
Specpure™
Ni in 2% HNO3
UN3264
Ni 10μg/mL
Liquid
MFCD00011137
100mL
Videos
SDS