missing translation for 'onlineSavingsMsg'
Learn More

Nickel naphthenate, Ni 5-12%, Thermo Scientific Chemicals

Catalog Number 15461537
Click to view available options
Quantity:
1 kg

 

Applied as a rubber catalyst. The product is a aid siccative and also acts as a stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 61788-71-4
Color Green
Physical Form Viscous Liquid
Quantity 1 kg
Molecular Formula C22H14NiO4
MDL Number MFCD00147723
UN Number UN3082
Synonym nickel naphthenate, naphthenic acids, nickel salts, nickel 2+ bis 2-naphthoate, nickel ii naphthenate, approx in toluene ni
Solubility Information Insoluble in water.
InChI Key UIEKYBOPAVTZKW-UHFFFAOYSA-L
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)[O-].C1=CC=C2C=C(C=CC2=C1)C(=O)[O-].[Ni+2]
IUPAC Name naphthalene-2-carboxylate;nickel(2+)
Molecular Weight (g/mol) 401.043
PubChem CID 14647680
Concentration or Composition (by Analyte or Components) Ni 5 to 12%
Chemical Name or Material Nickel naphthenate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'