missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel Iron, sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Magnetron sputtering source

Brand:  Alfa Aesar™ 040886.KS

Code : NEW

Additional Details : Weight : 0.01000kg


Product Code. 15423745

  • £388.80 / Each


EU Stock 2
Add to basket


Specifications

Specifications

Ni:Fe; 81:19 wt%
1 each
99.95%
MFCD01741928
Nickel Iron, sputtering target
50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
(metals basis)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Nickel Iron, sputtering target

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS