missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel Iron Molybdenum flake, -200 mesh

Brand:  Alfa Aesar™ 045610.A1

Code : NEW

Additional Details : Weight : 1.00000kg Transport : UN number : 3089 Chem class : 1 Pack group : III

Quantity 1kg

 View more versions of this product

Product Code. 15422417

  • £161.10 / 1kg


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

Nickel Iron Molybdenum flake
MFCD09910537
UN3089

Specifications

Ni:Fe:Mo; 81:17:2 wt%
1kg
Flakes
UN3089
Nickel Iron Molybdenum flake
0.29g/mL
MFCD09910537
−200 Mesh
Videos
SDS