missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Catalog Number p-7040959
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
60871-84-3
C2F6NiO6S2
356.82
MFCD00673740
KVRSDIJOUNNFMZ-UHFFFAOYSA-L
nickel ii trifluoromethanesulfonate, nickel ii triflate, nickel 2+ ditriflate, trifluoromethanesulfonic acid nickel ii salt, nickel triflate, acmc-209mmw, ni otf 2, nickelous trifluoromethanesulfonate, nickel 2+ ion ditriflate
9820095
nickel(2+);trifluoromethanesulfonate
[Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
This item is not returnable. View return policy
60871-84-3
C2F6NiO6S2
356.82
MFCD00673740
KVRSDIJOUNNFMZ-UHFFFAOYSA-L
nickel ii trifluoromethanesulfonate, nickel ii triflate, nickel 2+ ditriflate, trifluoromethanesulfonic acid nickel ii salt, nickel triflate, acmc-209mmw, ni otf 2, nickelous trifluoromethanesulfonate, nickel 2+ ion ditriflate
9820095
nickel(2+);trifluoromethanesulfonate
[Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

It is used as a pharmaceutical intermediate and as OLED chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and as OLED chemicals.

Solubility
Partly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 100°C to 106°C
Linear Formula Ni(OSO2CF3)2
Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Partly soluble in water.
Formula Weight 356.83
Percent Purity 96%
Chemical Name or Material Nickel(II) trifluoromethanesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'