missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) tetrafluoroborate hexahydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
It is mainly used as an electroplating intermediate. It is also used in medical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Lump |
Quantity | 100 g |
UN Number | UN3077 |
Solubility Information | Soluble in water. |
IUPAC Name | nickel(2+) bis(tetrafluoroboranuide) hexahydrate |
Formula Weight | 340.40 (232.30 Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 1.5 g/mL |
Chemical Name or Material | Nickel (II) tetrafluoroborate hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'