missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) phthalocyanine, Thermo Scientific Chemicals

Catalog Number p-7050784
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
14055-02-8
C32H16N8Ni
571.23
MFCD00041954
LVIYYTJTOKJJOC-UHFFFAOYSA-N
nickel phthalocyanine, nickel ii phthalocyanine, phthalocyanine, ni, nickel ii phthalocyanine, dye content 85 %
2735100
nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
[Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
This item is not returnable. View return policy
14055-02-8
C32H16N8Ni
571.23
MFCD00041954
LVIYYTJTOKJJOC-UHFFFAOYSA-N
nickel phthalocyanine, nickel ii phthalocyanine, phthalocyanine, ni, nickel ii phthalocyanine, dye content 85 %
2735100
nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
[Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color Blue to Purple
Quantity 5 g
Formula Weight 571.24
Physical Form Powder
Chemical Name or Material Nickel(II) phthalocyanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'