missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) perchlorate hydrate, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059278
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
13637-71-3
Cl2NiO8
257.59
MFCD00149804
ZLQBNKOPBDZKDP-UHFFFAOYSA-L
nickel perchlorate, unii-bq8a24mezo, perchloric acid, nickel 2+ salt, nickel ii perchlorate, nickel 2+ diperchlorate, bq8a24mezo, perchloric acid, nickel 2+ salt 2:1, nickelous perchlorate, perchloric acid, nickel salt, nickel perchlorate ni clo4 2
26158
[Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
This item is not returnable. View return policy
13637-71-3
Cl2NiO8
257.59
MFCD00149804
ZLQBNKOPBDZKDP-UHFFFAOYSA-L
nickel perchlorate, unii-bq8a24mezo, perchloric acid, nickel 2+ salt, nickel ii perchlorate, nickel 2+ diperchlorate, bq8a24mezo, perchloric acid, nickel 2+ salt 2:1, nickelous perchlorate, perchloric acid, nickel salt, nickel perchlorate ni clo4 2
26158
[Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Applied as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 140°C
Color Green
Physical Form Crystalline
Quantity 10 g
Linear Formula Ni(ClO4)2·xH2O
UN Number UN1481
Solubility Information Very soluble in water. Soluble in many organic solvents.
IUPAC Name nickel(2+) diperchlorate
Formula Weight 257.59 (Anhydrous)
Percent Purity 99.998%
Odor Odorless
Refractive Index 1.55
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Nickel (II) perchlorate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'