missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, green, Ni 78.5%, Thermo Scientific Chemicals

Catalog Number 15402157
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
1313-99-1
NiO
74.69
MFCD00011145
GNRSAWUEBMWBQH-UHFFFAOYSA-N
nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
14805
oxonickel
O=[Ni]
This item is not returnable. View return policy
1313-99-1
NiO
74.69
MFCD00011145
GNRSAWUEBMWBQH-UHFFFAOYSA-N
nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
14805
oxonickel
O=[Ni]

Nickel(II) oxide is used in the preparation of nickel alloys including nickel steel alloys, manufacture of glass and porcelain paints. Further nickel oxide is the main component in nickel-iron battery (Edison battery), nickel-cadmium rechargeable battery, and also used in ceramic industry for making frits and porcelain glazes. It also acts as a hydrogenation catalyst. NiO/CNTs (nickel oxide/carbon nanotubes) could be a potential cathode catalyst for oxygen reduction reaction (ORR) in microbial fuel cells (MFCs). It is blended with other high purity oxides and used in a variety of semiconductor applications such as thermistors and varistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1984°C
Color Green
Physical Form Powder
Quantity 250 g
Assay Percent Range Ni 78.5%
Solubility Information Soluble in ammonium hydroxide. Insoluble in water.
Formula Weight 74.71
Odor Odorless
Density 6.67
Chemical Name or Material Nickel(II) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'