missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) oxide, black, Ni 76%, (contains Ni{2}O{3}), Thermo Scientific Chemicals
45.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Description
Nickel(II) oxide is used in fuel cell electrodes, storage batteries, electronic circuit boards, electrical ceramics, and as a pigment for ceramics and glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1984°C |
Color | Black |
Physical Form | Powder |
Quantity | 250 g |
Merck Index | 14,6512 |
Solubility Information | Soluble in acids. Insoluble in water. |
Formula Weight | 74.71 |
Concentration or Composition (by Analyte or Components) | Ni 76%, (contains Ni2O3) |
Odor | Odorless |
Density | 6.67 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'