missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel(II) oxide, 99.995% (metals basis)

Vacuum deposition

Brand:  Alfa Aesar™ 044423.22

Code : NEW

Additional Details : CAS Number : 1313-99-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11450710

  • £438.90 / 100g


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Specifications

Chemical Identifiers

Nickel (II) oxide
NiO
MFCD00011145
1313-99-1
74.71
14,6512

Specifications

Nickel (II) oxide
Pieces
(metals basis)
74.71
14,6512
Odorless
Green to black
100g
1313-99-1
99.995%
NiO
MFCD00011145
6.67g/mL
3 to 12mm
1984°C
Videos
SDS