missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Nitrate Hexahydrate, purum p.a, ≥97.0% (KT), Honeywell™

Catalog Number p-7120621
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
13478-00-7
H12N2NiO12
290.79
MFCD00149805
AOPCKOPZYFFEDA-UHFFFAOYSA-N
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
61630
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
This item is not returnable. View return policy
13478-00-7
H12N2NiO12
290.79
MFCD00149805
AOPCKOPZYFFEDA-UHFFFAOYSA-N
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
61630
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Purum p.a., crystallized,≥97.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Melting Point 56.7°C
Color Blue Green
pH 4.0 (at 20°C)
Physical Form Solid
Quantity 250 g
Linear Formula Ni(NO3)2 · 6H2O
UN Number UN2725
Solubility Information 2.385g/L at 20°C
IUPAC Name nickel(2+) hexahydrate dinitrate
Formula Weight 290.79g/mol
Percent Purity ≥97.0% (KT)
Packaging Poly bottle
Density 2.05g/mL at 25°C
Chemical Name or Material Nickel(II) nitrate hexahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'