missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) formate dihydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Nickel(II) formate dihydrate is used in nickel catalyst preparations. It is also used as a biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 130°C to 140°C-2H2O |
Color | Green |
Physical Form | Powder |
Quantity | 100 g |
Solubility Information | In the range 180-200° decomposes to Ni,CO,CO2,H2,H2O,CH4. Partially soluble in water |
Formula Weight | 184.77 (148.74 Anhydrous) |
Appearance | Green powder |
Decomposition Information | In the range 180° to 200°C decomposes to Ni,CO,CO2 |
Density | 2.15 g/cm3 |
Chemical Name or Material | Nickel(II) formate dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'